Order Entry
Sweden
ContactUsLinkComponent
 
 
Tricaine methanesulphonate, powder
 
 
Tricaine methanesulphonate, powder

 

  • Identity Test:
    Passes
  • FTIR:
    Conforms to Standard
  • Melting Point :
    145 - 150°C
  • pH (1% aq soln):
    <7
  • Loss on Drying:
    ≤5%