Order Entry
Sweden
ContactUsLinkComponent
1,4,5,8-Tetraamino-9,10-anthraquinone 30% (dye content), Sigma-Aldrich®
  215643-5G
 :  
undefined
1,4,5,8-Tetraamino-9,10-anthraquinone 30% (dye content), Sigma-Aldrich®
  215643-5G
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
  • Appearance:
    Grey to black powder
  • Infrared spectrum:
    Conforms to structure
  • UV-visible spectrum:
    0.01 g/L in 50% ethanol w/1 drop 1N NaOH
 :
 :268.27 g/mol
 :332 °C
 :219-603-7
 :GHS05
 :GHS07
 :GHS08
 :Room Temperature
 :2475-45-8
 :C₁₄H₁₂N₄O₂
 :MFCD00001225