Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pyridino[3,2-b]pyrazine
Pyridino[3,2-b]pyrazine ≥98%
Pyridino[3,2-b]pyrazine ≥98%
Irritant
Pyridino[3,2-b]pyrazine ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
Pyridino[3,2-b]pyrazine ≥98%
Supplier:
Thermo Fisher Scientific
About this item
CAS Number:
322-46-3
:
Formula:
C₇H₅N₃
MDL Number:
MFCD00006717
:
131.14 g/mol
Melting Pt:
142…144 °C
EINECS:
206-294-9
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFAB22843
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation