Benzyl butyl phthalate-[ring-D4] 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA16168010CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:93951-88-3
- Description:Benzyl butyl phthalate-[ring-D4] 100 µg/ml in cyclohexane
- Formula:C₁₉₂H₄H₁₆O₄
- Description:Benzyl butyl phthalate-[ring-D4]