Order Entry
Sweden
ContactUsLinkComponent
SPE columns, CHROMABOND® C18 f (fast flow)
SPE columns, CHROMABOND® C18 f (fast flow)
Leverantör:  MACHEREY-NAGEL
Certifikat
undefined
SPE columns, CHROMABOND® C18 f (fast flow)
Leverantör:  MACHEREY-NAGEL
Specifikationer
  • Catalog No:
  • MANA730403
  • MANA730009
  • 554-6595
  • MANA730402
  • Carbon load:
  • 14%
  • 14%
  • 14%
  • 14%
  • Endcapped:
  • No, No
  • No, No
  • No, No
  • No, No
  • pH range:
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • Surface area:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
Specifikationer
Frequently Bought Together