Order Entry
Portugal
ContactUsLinkComponent
CloneR™-2 Supplement
CloneR™-2 Supplement
Catalog # STMC100-0691
CAS Number:  
undefined
CloneR™-2 Supplement
Catalog # STMC100-0691
CAS Number:  
Specifications
  • Und.:
    25 mL
  • Descrição:
    CloneR™ 2 Supplement
Specifications
About this item
Defined supplement for improving survival of human ES and iPS cells in single-cell workflows.
Generate clonal human pluripotent stem cell (hPSC) lines that maintain their genomic integrity and downstream differentiation potential with this supplement. Using CloneR™2 will increase the cloning efficiency and survival of human embryonic stem (ES) and induced pluripotent stem (iPS) cells under high-stress conditions, including seeding at low or high densities. For gene-editing workflows, add CloneR™2 to improve ES and iPS cell survival following electroporation and during clonal deposition. It also facilitates the survival and expansion of clonal lines generated by low-density seeding (<25 cells/cm²) or by FACS-sorting cells at 1 cell/well, without requiring single-cell adaptation. Also use CloneR™2 supplement to increase survival of single cells plated at higher densities, including under stressful conditions such as the post-thaw recovery of ES and iPS cell lines and when creating monolayers ahead of downstream differentiation. Defined and serum-free, CloneR™2 supplement is compatible for use with TeSR™ ES and iPS cell maintenance media, as well as a choice of cell culture matrix.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').