Order Entry
Portugal
ContactUsLinkComponent
Precision density hydrometer, range: 1.500-1.600 in 0.001 g/cm , total length: 300 mm, body diameter: 21 mm, without thermometer, adjusting temperature: 20°C, including works-calibration certificate with 3 calibration points 1 * 1 items
Catalog # SCNL3031010/03/3P
New Product
Precision density hydrometer, range: 1.500-1.600 in 0.001 g/cm , total length: 300 mm, body diameter: 21 mm, without thermometer, adjusting temperature: 20°C, including works-calibration certificate with 3 calibration points 1 * 1 items
Catalog # SCNL3031010/03/3P