Eppendorf Conical Tubes 15 mL,BioBased, sterile, pyrogen, DNase, RNase, and DNA free, 400 pcs., 8 packs of 50 Tubes each 1 * 400 items
Catalog # EPPE0030122.631
Supplier: EPPENDORF
New Product
Eppendorf Conical Tubes 15 mL,BioBased, sterile, pyrogen, DNase, RNase, and DNA free, 400 pcs., 8 packs of 50 Tubes each 1 * 400 items
Catalog # EPPE0030122.631
Supplier: EPPENDORF
New Product
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK