Order Entry
Portugal
ContactUsLinkComponent
Human Liver Arginase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Liver Arginase in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 96 Tests
Catalog # ANTIA2226-96
Supplier:  ANTIBODIES.COM
New Product
Human Liver Arginase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Liver Arginase in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 96 Tests
Catalog # ANTIA2226-96
Supplier:  ANTIBODIES.COM