Order Entry
Portugal
ContactUsLinkComponent
Human Growth Hormone ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Growth Hormone in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 48 Tests
Catalog # ANTIA1709-48
Supplier:  ANTIBODIES.COM
New Product
Human Growth Hormone ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of human Growth Hormone in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other 1 * 48 Tests
Catalog # ANTIA1709-48
Supplier:  ANTIBODIES.COM