Order Entry
Portugal
ContactUsLinkComponent
Consists of amino acids Arg319-Phe541 of SARS-CoV-2 Spike Glycoprotein RBD (Alpha B.1.1.7 Variant) containing the following mutation : N501Y. The human Fc tag consists of amino acids Glu99-Ala330. 1 * 50 µG
Catalog # ANTIA318195-50
Supplier:  ANTIBODIES.COM
New Product
Consists of amino acids Arg319-Phe541 of SARS-CoV-2 Spike Glycoprotein RBD (Alpha B.1.1.7 Variant) containing the following mutation : N501Y. The human Fc tag consists of amino acids Glu99-Ala330. 1 * 50 µG
Catalog # ANTIA318195-50
Supplier:  ANTIBODIES.COM