Order Entry
Portugal
ContactUsLinkComponent
15-Deoxy-delta 12, 14-PGJ2 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative deter mination of 15-Deoxy-delta 12, 14-PGJ2 in serum, plasma, and other biological fluids. 1 * 96 Tests
Catalog # ANTIA326984-96
Supplier:  ANTIBODIES.COM
New Product
15-Deoxy-delta 12, 14-PGJ2 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative deter mination of 15-Deoxy-delta 12, 14-PGJ2 in serum, plasma, and other biological fluids. 1 * 96 Tests
Catalog # ANTIA326984-96
Supplier:  ANTIBODIES.COM