Order Entry
Portugal
Orders LinkContactUsLinkComponent
Mixing Systems, SpinBag

Mixing Systems, SpinBag

SpinBag mixing systems, 50 L
Catalog # PARKPHSBU050LGI
Supplier:   PARKER HANNIFIN
Mixing Systems, SpinBag
Catalog # PARKPHSBU050LGI
Supplier:  PARKER HANNIFIN
Mixing Systems, SpinBag

SpinBag mixing systems, 50 L

Price
825,00 €
Log in for availability
1 * 1 SET
Product Details & Documents

Parker's compact SpinBag Mixing System offers the advantages of single-use in a bench top mixing solution.

  • Constructed of fully validated DuraPure C93 film offering added seal strength, low gas permeability and optical clarity which can be heated to 60 °C
  • Stores flat and ideal for preparation of cell culture media, buffers, bioreactor feeds, and suspension/re-suspension applications
  • Easily integrated with filters, sensors and pumping systems
  • Available with standard configurations, or a variety of ports and tubing options in non-irradiated or gamma irradiated
  • Platinum-cured silicone coated magnetic stir bar offers optimal mixing performance and eliminates the need to validate another material
  • Manufactured in an ISO Class 7 cleanroom

Parker's 3D SpinBag bioprocess containers are constructed of durable, particle-free, DuraPure C93 film, each containing a platinum-cured silicone coated magnetic spin bar. When placed on a mixing plate, the spin bar rotates to create a vortex of motion for thorough mixing/stirring of fluid contents. Parker SpinBags are available in a variety of sizes and port configurations to easily integrate within your application.

Specifications
  • Description:
    SpinBag mixing systems, 50 L
  • Seq:
    02
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • SpinBag housing only, 50 L, polycarbonate
    SpinBag housing only, 50 L, polycarbonate
    PARKPHSBH050L
    Log in for availability
    1 * 1 SET - 783,00 €

Product Information

  • Catalog #
    Availability
    Price
  • SpinBag housing only, 15 L, polycarbonate
    SpinBag housing only, 15 L, polycarbonate
    PARKPHSBH015L
    Log in for availability
    1 * 1 SET - 653,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.