Order Entry
Portugal
ContactUsLinkComponent
PRIME-XV NK Cell CDM is the first commercially-available, chemically defined, and animal component-free medium for the ex vivo expansion of NK and NKT cells. Designed for use in NK and NKT cell-based immunotherapy research and translational applicati 1 * 1 L
Catalog # SHBT91215
New Product
PRIME-XV NK Cell CDM is the first commercially-available, chemically defined, and animal component-free medium for the ex vivo expansion of NK and NKT cells. Designed for use in NK and NKT cell-based immunotherapy research and translational applicati 1 * 1 L
Catalog # SHBT91215