PRIME-XV Tumorsphere SFM is a serum-free medium optimized for the tumorsphere formation of cancer initiating/ stem cells. This product does not contain antibiotics. Designed and manufactured to facilitate transfer from research to clinic:FDA, Federal 1 * 100 mL
Catalog # SHBT91130
Supplier: Shenandoah Biotechnology