Order Entry
Portugal
Orders LinkContactUsLinkComponent
Human Recombinant LIF, ACF

Human Recombinant LIF, ACF

1.000 µG
Catalog # STMC78149.2
Marketsource
Human Recombinant LIF, ACF
Catalog # STMC78149.2
Marketsource
CAS Number:  
Human Recombinant LIF, ACF
Human Recombinant LIF, ACF

1.000 µG

CAS Number:  
Price
4 490,00 €
Log in for availability
1 * 1.000 µG
Product Details & Documents

Leukemia inhibitory factor (LIF) is an interleukin 6 class cytokine that regulates a broad variety of developmental functions. After LIF binds to LIF receptor (LIFR), LIFR associates with gp130 and activates JAK/STAT and MAPK signaling (Auernhammer and Melmed; Suman et al.). LIFR activation of STAT3 is essential for maintaining the mouse embryonic stem cell phenotype (Niwa et al.). Produced by the endometrium, LIF plays an important autocrine and paracrine role in implantation by regulating proliferation, invasion, and differentiation of trophoblasts following blastocyst attachment (Auernhammer and Melmed; Suman et al.). Human LIF can be used for the maintenance of mouse embryonic stem cells, however mouse LIF cannot bind to the human receptor, thus rendering mouse LIF inactive (Dahéron et al.). LIF is produced by CD4+ and activated regulatory T cells, and promotes Foxp3 expression, while repressing Th17 lineage-specific genes (Metcalfe). LIF is also secreted by mesenchymal stromal cells, where it supports hematopoiesis and immune modulation (Nasef et al.). This product is animal component-free.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    1.000 µG
  • Cat. No.:
    STMC78149.2
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    LIF, ACF
  • Purity:
    ≥95%
  • Species:
    Human

Specifications