Order Entry
Portugal
Orders LinkContactUsLinkComponent
Human Recombinant IGF-II

Human Recombinant IGF-II

1.000 µG
Catalog # STMC78023.2
Marketsource
Human Recombinant IGF-II
Catalog # STMC78023.2
Marketsource
CAS Number:  
Human Recombinant IGF-II
Human Recombinant IGF-II

1.000 µG

CAS Number:  
Price
2 020,00 €
Log in for availability
1 * 1.000 µG
Product Details & Documents

Insulin-like growth factor 2 (IGF-II) is a polypeptide that belongs to the family of insulin-like growth factors that are similar in molecular structure to proinsulin. IGF-II binding to IGF-I receptor activates a signaling cascade via the MAPK and PI3K pathways. IGF-II affects growth, differentiation, and survival of cells in a variety of tissues during embryonic development. It stimulates proliferation and migration of primary lymphatic endothelial cells, and induces lymphangiogenesis (Guvakova).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    1.000 µG
  • Cat. No.:
    STMC78023.2
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IGF-II
  • Protein synonyms:
    Insulin-like growth factor II|Insulin-like growth factor type 2|Somatomedin A|T3M-11-derived growth factor
  • Purity:
    ≥95%
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.