Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Portugal
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FLUORESCEIN RECOMBINANT MONOCLONAL ANTIBODY (4-4-20 ENHANCED) (HUMAN IGG3Κ) 1 * 200 µG
Catalog # ENZOENZABS5070200
Supplier:
ENZO LIFE SCIENCES
Expand image
FLUORESCEIN RECOMBINANT MONOCLONAL ANTIBODY (4-4-20 ENHANCED) (HUMAN IGG3Κ) 1 * 200 µG
Catalog # ENZOENZABS5070200
Supplier:
ENZO LIFE SCIENCES
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Transfer pipettes
Supplier: RATIOLAB
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF