SONIDEGIB DIPHOSPHATE 1 * 200 mg
Catalog # ENZOENZCHM2370200
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
Aluminium ≥90% stabilised, fine powder, Supelco®
Supplier: Merck
Hydrazinium sulphate, GR for analysis, Supelco®
Supplier: Merck
Sodium hexanitritocobaltate(III), AnalaR NORMAPUR® ACS, Reag. Ph. Eur.
Supplier: VWR Chemicals
Sodium carbonate ≥99.9%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium sulphate, anhydrous 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Ammonium formate ≥97%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ammonium sulphide 20% in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Calcium nitrate tetrahydrate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ammonium acetate ≥98.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Silver nitrate ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium hydroxide 98-100.5%, pellets, Puriss., meets analytical specification of BP, NF, Ph. Eur., E524, Fluka™
Supplier: Honeywell Chemicals
Lithium hydroxide monohydrate ≥98.5%, Purum, Fluka™
Supplier: Honeywell Chemicals
Sodium diatrizoate 99%
Supplier: Thermo Fisher Scientific
Benzyl alcohol ACS
Supplier: Thermo Fisher Scientific
Ruthenium(III) chloride hydrate 97%
Supplier: Apollo Scientific