Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Portugal
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PROTEASOME 20S Α2 SUBUNIT MONOCLONAL ANTIBODY (MCP21) (AGAROSE IMMOBILIZED) 1 * 0,5 mL
Catalog # ENZOBMLPW83350500
Supplier:
ENZO LIFE SCIENCES
Expand image
PROTEASOME 20S Α2 SUBUNIT MONOCLONAL ANTIBODY (MCP21) (AGAROSE IMMOBILIZED) 1 * 0,5 mL
Catalog # ENZOBMLPW83350500
Supplier:
ENZO LIFE SCIENCES
Frequently Bought Together
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection