Order Entry
Portugal
Orders LinkContactUsLinkComponent
Streptozocin ≥97%

Streptozocin ≥97%

Glass bottle for solids, 100 mg
Catalog # ENZOALX3800105100
Health
Supplier:   Enzo Life Sciences
Streptozocin ≥97%
Catalog # ENZOALX3800105100
Supplier:  Enzo Life Sciences
CAS Number:  18883-66-4
Streptozocin ≥97%
Streptozocin ≥97%

Glass bottle for solids, 100 mg

CAS Number:  18883-66-4
Price
139,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

Widely used diabetes inducer in rodents. Inhibition of β-cell O-GlcNAcase by streptozotocin is the mechanism that accounts for its diabetogenic toxicity. N-nitroso-containing antibiotic, acting as a nitric oxide (NO) donor and vasorelaxant. Relaxes phenylephrine-contracted aortic rings at 10µM. Potent methylating agent for DNA.

  • O-GlcNAcase inhibitor

White to pale yellow crystalline solid.

Soluble in water (nH2O at 102.8mg/ml) or 100% ethanol (200 proof at 0.92mg/ml).

Synonyms:  1-Methyl-1-nitroso-3-((2S,3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)urea, Streptozotocin
  • CAS Number:18883-66-4
  • Formula:C₈H₁₅N₃O₇
  • MDL Number:MFCD00006607
  • Molecular Weight:265.2 g/mol
  • Storage Temperature:Freezer
  • EINECS:242-646-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications