Gentamicin sulphate
Glass bottle for solids, 25 g
Catalog # ENZOALX380003G025
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
- CAS Number:1405-41-0
- MDL Number:MFCD00270181
- Merck Index:13,04403
- Melting Pt:218…237 °C
- Storage Temperature:Ambient
- EINECS:215-778-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



