Order Entry
Portugal
Orders LinkContactUsLinkComponent
Gentamicin sulphate

Gentamicin sulphate

Glass bottle for solids, 1 g
Catalog # ENZOALX380003G001
Health
Supplier:   Enzo Life Sciences
Gentamicin sulphate
Catalog # ENZOALX380003G001
Supplier:  Enzo Life Sciences
CAS Number:  1405-41-0
Gentamicin sulphate

Glass bottle for solids, 1 g

CAS Number:  1405-41-0
Price
85,80 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
  • CAS Number:1405-41-0
  • MDL Number:MFCD00270181
  • Merck Index:13,04403
  • Melting Pt:218…237 °C
  • Storage Temperature:Ambient
  • EINECS:215-778-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.