Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Portugal
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
7 components: 100mg/l each of Fe Al Cu Zn Ba Mn Cr in HNO3 1 * 50 mL
Catalog # CMPLRM093370L05
Supplier:
CUSTOM MADE CHEMICALS LAB
Expand image
7 components: 100mg/l each of Fe Al Cu Zn Ba Mn Cr in HNO3 1 * 50 mL
Catalog # CMPLRM093370L05
Supplier:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
Test strips, glucose, Reflectoquant®, Supelco®
Supplier: Merck
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Reagents for chlorine determination using Lovibond® MD photometers
Supplier: Lovibond
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
COD photometric test kits, Test’n Tube
Supplier: Hach
Reagent powder pillows for determination of molybdenum, MolyVer®
Supplier: Hach