Order Entry
Portugal
Orders LinkContactUsLinkComponent
Test kits, COD cuvette tests for salt water
Test kits, COD cuvette tests for salt water
COD cuvette test (LCK) for salt water, 70 - 250 mg/L, Dichromate, 376 g
Catalog # HACHLCK1714
Supplier:   Hach
Test kits, COD cuvette tests for salt water
Catalog # HACHLCK1714
Supplier:  Hach
CAS Number:  
Test kits, COD cuvette tests for salt water
Test kits, COD cuvette tests for salt water
COD cuvette test (LCK) for salt water, 70 - 250 mg/L, Dichromate, 376 g
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 25 Tests
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Applicable for COD testing in water samples with high chloride concentrations up to 20000 mg/L.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
  • No cumbersome sample pre-treatment required
Pre-dosed reagents for maximum safety. Easy to use and photometric evaluation, packed in a fully equipped box.
Specifications
Specifications
  • Chemical method:
    Dichromate
  • Pk:
    376 g
  • Description:
    COD cuvette test (LCK) for salt water
  • Range:
    70 - 250 mg/L