Syringe Filter, Nylon, 0.22 um, 30 mm 1 * 30 items
STMC200-0589
Frequently Bought Together
Filter plates, 96-well, for solvent filtration, AcroPrep™ Advance
: Cytiva (Formerly Pall Lab)
Mixed cellulose ester membranes, non-sterile, 0,6 µm, Whatman™
: Whatman products (Cytiva)
Centrifugal filtration units, Vivaspin® 500
: Sartorius
Dialysis membrane tubing spectra/Por® 7
: Spectrum Laboratories
Qualitative filter papers, high-purity
: Sartorius
Membrane filters, type 111
: Sartorius
Filtration capsules, Sartopore® 2 XLG
: Sartorius
General purpose qualitative filter papers
: Ahlstrom-Munksjö