Order Entry
Portugal
ContactUsLinkComponent
Special program to test steam penetration to pores and hollow loads. The cycle includes 3.5 minutes sterilization after pre-vacuum phase 1 * 1 items
Catalog # TUTTADD222-0501
Supplier:  TUTTNAUER
Special program to test steam penetration to pores and hollow loads. The cycle includes 3.5 minutes sterilization after pre-vacuum phase 1 * 1 items
Catalog # TUTTADD222-0501
Supplier:  TUTTNAUER
Frequently Bought Together