0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Portugal
ContactUsLinkComponent
3-METHOXY-P-TYRAMINE-D3 HYDROCHLORIDE 1 * 10 mg
PROOTRCM332322-10
:
LGC Standards PROMOCHEM
3-METHOXY-P-TYRAMINE-D3 HYDROCHLORIDE 1 * 10 mg
PROOTRCM332322-10
:
LGC Standards PROMOCHEM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
Buffer solutions in sachets, Certipur®, Supelco®
: Merck
VWR®, Conductivity standard solutions
: VWR Chemicals
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
: Merck
pH buffers, ready to use
: WTW