Reagecon System Suitability Set to USP for Total Organic Carbon (TOC) suitable for use with Sievers 900/M9 Analysers, 3x40ml 1 * 3 items
Catalog # REGERTOCLWUSP1
Supplier: REAGECON
Reagecon System Suitability Set to USP for Total Organic Carbon (TOC) suitable for use with Sievers 900/M9 Analysers, 3x40ml 1 * 3 items
Catalog # REGERTOCLWUSP1
Supplier: REAGECON
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Calcium carbonate, Certipur®, Supelco®
Supplier: Merck
Tin(II) chloride 30% in hydrochloric acid 20% Reag. Ph. Eur. 1085001
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Redox (ORP) Standards
Supplier: VWR Chemicals
Methyl stearate
Supplier: EHRENSTORFER
Indium Standard for ICP 1 * 100 mL
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
Polysorbate, Reference Standards
Supplier: USP
pH buffer solutions
Supplier: Hach