Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Portugal
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Canine CD19, His Tag (CD9-C52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Gly 297 (Accession # XP_0385246 89.1). 1 * 100 µG
Catalog # ACRBCD9-C52H4100UG
Supplier:
ACROBIOSYSTEMS
Expand image
Canine CD19, His Tag (CD9-C52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Gly 297 (Accession # XP_0385246 89.1). 1 * 100 µG
Catalog # ACRBCD9-C52H4100UG
Supplier:
ACROBIOSYSTEMS
Frequently Bought Together
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR®, Microcentrifuge Tubes and Screw Caps
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Transfer Pipettes, Graduated
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Multiwell cell culture plates, Falcon®
Supplier: Corning
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection