Masterflex® L/S® Replacement Fuse, Standard. 1.5 A/250 V, 3 AG
Catalog # MFLX77500-12
Supplier: Avantor Fluid Handling
About this item
Make the most of your drive with these replacement parts.
- Optimize performance of your Masterflex pumps
Suitable for use with Masterflex® B/T® drive 77111-30.
For use with the following Masterflex L/S drive and controller: 07552-75 and 07552-76.
- Description:Replacement Fuse, 1,5 A/250 V, 3 AG, standard
- Environmentally Preferable:
- Compatible with:Masterflex L/S Drives
- Pk:2
Frequently Bought Together