Order Entry
Portugal
Orders LinkContactUsLinkComponent
 

PQ401

50 mg
Catalog # TCIAP2479-50MG
Supplier:   TCI
PQ401
Catalog # TCIAP2479-50MG
Supplier:  TCI
CAS Number:  196868-63-0
CAS Number:  196868-63-0
Price
289,00 €
Log in for availability
1 * 50 mg
Product Details & Documents
Synonyms:  1-(5-Chloro-2-methoxyphenyl)-3-(2-methylquinolin-4-yl)urea, N-(5-Chloro-2-methoxyphenyl)-N’-(2-methyl-4-quinolinyl)urea, IGF-IR Inhibitor
  • CAS Number:196868-63-0
  • Formula:C₁₈H₁₆ClN₃O₂
  • MDL Number:MFCD00160558
  • Molecular Weight:341.8 g/mol
Specifications

Specifications

  • Pk:
    50 mg

Specifications