Order Entry
Portugal
Orders LinkContactUsLinkComponent
 

PQ401

10 mg
Catalog # TCIAP2479-10MG
Supplier:   TCI
PQ401
Catalog # TCIAP2479-10MG
Supplier:  TCI
CAS Number:  196868-63-0
CAS Number:  196868-63-0
Price
90,00 €
Log in for availability
1 * 10 mg
Product Details & Documents
Synonyms:  1-(5-Chloro-2-methoxyphenyl)-3-(2-methylquinolin-4-yl)urea, N-(5-Chloro-2-methoxyphenyl)-N’-(2-methyl-4-quinolinyl)urea, IGF-IR Inhibitor
  • CAS Number:196868-63-0
  • Formula:C₁₈H₁₆ClN₃O₂
  • MDL Number:MFCD00160558
  • Molecular Weight:341.8 g/mol
Specifications

Specifications

  • Pk:
    10 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.