Order Entry
Portugal
ContactUsLinkComponent
The PowerPlex Fusion 6C System is a 6-color, 27-locus multiplex for human identification applications including forensic analysis, r elationship testing and research use. Use with extracted DNA samples, punches and pretreated swabs. 1 * 800 Assays
Catalog # PROMDC2780
Supplier:  Promega
undefined
The PowerPlex Fusion 6C System is a 6-color, 27-locus multiplex for human identification applications including forensic analysis, r elationship testing and research use. Use with extracted DNA samples, punches and pretreated swabs. 1 * 800 Assays
Catalog # PROMDC2780
Supplier:  Promega
Frequently Bought Together