Order Entry
Portugal
Orders LinkContactUsLinkComponent
VWR® Septa ND8

VWR® Septa ND8

Red rubber/beige PTFE, 45° shore A, 1,0 mm
Catalog # 548-1460A
Supplier:   VWR Collection
VWR® Septa ND8
Catalog # 548-1460A
Supplier:  VWR Collection
VWR® Septa ND8
VWR® Septa ND8

Red rubber/beige PTFE, 45° shore A, 1,0 mm

Price
184,00 €
Log in for availability
1 * 1.000 unid.
Product Details & Documents

Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.

  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available

Septa choice depends on the application.

Specifications
  • Hardness:
    45° shore A
  • Septum:
    Red rubber/beige PTFE
  • Seq:
    07
  • Thickness:
    1,0 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.