Order Entry
Portugal
Orders LinkContactUsLinkComponent
Fluorescamine

Fluorescamine

Bottle, 5 g
Catalog # APOSBIF4008-5G
Irritant
Supplier:   Apollo Scientific
Fluorescamine
Catalog # APOSBIF4008-5G
Supplier:  Apollo Scientific
CAS Number:  38183-12-9
Fluorescamine

Bottle, 5 g

CAS Number:  38183-12-9
Price
1 100,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Reacts with primary amines to form stable, highlyfluorescent products.

Synonyms:  Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione
  • CAS Number:38183-12-9
  • Formula:C₁₇H₁₀O₄
  • MDL Number:MFCD00005928
  • Molecular Weight:278.26 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.