Order Entry
Portugal
ContactUsLinkComponent
Human IL-31, His Tag IL1-H5247 is expressed from human 293 cells HEK293. It contains AA Ser 24 - Thr 164 Accession Q6EBC2-1. 1 * 100 µG
Catalog # ACRBIL1-H5247100UG
Supplier:  ACROBIOSYSTEMS
Human IL-31, His Tag IL1-H5247 is expressed from human 293 cells HEK293. It contains AA Ser 24 - Thr 164 Accession Q6EBC2-1. 1 * 100 µG
Catalog # ACRBIL1-H5247100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together