SUB-X CLEARING MEDIUM 1 * 5 L
Catalog # LEIH3803672E
Supplier: LEICA HISTOLOGY
Frequently Bought Together
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Supplier: VWR Chemicals
Dihydroethidium ≥95%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Safranine O, Certifiable Grade
Supplier: VWR Chemicals
Reagents for ammonia determination using Lovibond® MD photometers
Supplier: Lovibond
Nitrite VHR L, 500 ml
Supplier: Lovibond
Sodium Hydroxide solution 32 % w/w for nitrogen determination 1 * 25 L
Supplier: PanReac AppliChem