Order Entry
Portugal
Orders LinkContactUsLinkComponent
Horizontal fold flat disposable masks, Springfit™
Horizontal fold flat disposable masks, Springfit™
Springfit™ Typhoon™ Disposable mask, valved, FFP3
Catalog # JSPABGA182-206-000
Supplier:   JSP
Horizontal fold flat disposable masks, Springfit™
Catalog # JSPABGA182-206-000
Supplier:  JSP
Horizontal fold flat disposable masks, Springfit™
Horizontal fold flat disposable masks, Springfit™
Springfit™ Typhoon™ Disposable mask, valved, FFP3
Price
49,10 €
Log in for availability
1 * 10 unid.
Product Details & Documents
The SpringFit™ fold flat disposable mask achieves an unrivalled secure fit with the use of a unique endoskeleton structure. SpringFit™ introduces an abundance of features that maximise fit, comfort, compatibility, and performance.
  • Fold flat disposable mask
  • Compact and easily pocketed
  • Unique endoskeleton structure
  • Polypropylene inner skeleton
  • Available in FFP3 and FFP2, valved and unvalved
The SpringFit™ fold flat disposable mask is available in FFP3 and FFP2, valved and unvalved, with a nuisance odour carbon layer available on the FFP3 model. The Unique Typhoon™ valve has been designed to reduce breathing resistance, heat, and moisture build-up. The valve is unaffected by any moisture, and therefore can be used with the same performance under very low temperatures.
The fold flat design makes SpringFit™ more compact, and easily pocketed. SpringFit™ is also individually packed to ensure a high level of hygiene.
Certifications: EN149:2001+A1:2009.
Specifications
  • Description:
    Springfit™ Typhoon™ Disposable mask, valved
  • Seq:
    04
  • Class:
    FFP3