Order Entry
Portugal
ContactUsLinkComponent
Cyanide Distillation Apparatus 3350-C, 500 Ml Distilling apparatus to determine soluble and insoluble cyanides in water Apparatus meets the requirements of E.P.A. SW-846 methods 9010 and 9012, and ASTM method D-2036. 1 * 1 items
Catalog # BIBBS83350-C
Supplier:  SciLabware
Cyanide Distillation Apparatus 3350-C, 500 Ml Distilling apparatus to determine soluble and insoluble cyanides in water Apparatus meets the requirements of E.P.A. SW-846 methods 9010 and 9012, and ASTM method D-2036. 1 * 1 items
Catalog # BIBBS83350-C
Supplier:  SciLabware
Frequently Bought Together