Order Entry
Portugal
ContactUsLinkComponent
SERPING1 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human Serpin G1 in cell cul ture supernates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5461
Supplier:  Abnova
SERPING1 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human Serpin G1 in cell cul ture supernates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # ABNOKA5461
Supplier:  Abnova
Frequently Bought Together