Order Entry
Portugal
Orders LinkContactUsLinkComponent
Microplate readers, multi-mode, SpectraMax® iD3

Microplate readers, multi-mode, SpectraMax® iD3

SpectraMax® iD3 multi-mode microplate reader with injectors
Catalog # 735-0390
Supplier:   MOLECULAR DEVICES
Microplate readers, multi-mode, SpectraMax® iD3
Catalog # 735-0390
Supplier:  MOLECULAR DEVICES
Microplate readers, multi-mode, SpectraMax® iD3
Microplate readers, multi-mode, SpectraMax® iD3

SpectraMax® iD3 multi-mode microplate reader with injectors

Price
Inquire For Price
Log in for availability
1 * 1 unid.
Product Details & Documents

The SpectraMax® iD3 multi-mode microplate reader measures absorbance, fluorescence and luminescence and is ideal for busy multi-user, multi-application labs. Large, high resolution touchscreen interface allows you to easily set up methods, run experiments or view tutorial videos without the need for a dedicated computer workstation.

  • Easily and quickly pull up your custom protocols with a single tap, and reduce the possibility of confusion in busy, multi-user labs, with built-in near-field communication (NFC) functionality
  • Access your results quickly, with no need to physically go to the instrument, with QuickSync
  • Comes with SoftMax® Pro Acquisition and Analysis software – the world’s most published microplate reader control and data analysis software
  • Working in a regulated GMP/GLP environment? Validate your instrument and software with an extensive suite of GxP tools and services
Specifications
  • Description:
    SpectraMax® iD3 multi-mode microplate reader with injectors
  • Seq:
    02
  • Incubation range:
    5 °C above ambient to 65 °C
  • Light source:
    Xenon flash lamp
  • Temperature uniformity:
    ±0,75 °C
  • Photometric range:
    0 - 4,0 OD
  • Shaking:
    Orbital and infinity
  • Wavelength bandwidth:
    4,0 nm
  • Temperature accuracy:
    ±1 °C @ 37 °C
  • Wavelength accuracy:
    ±2,0 nm
  • Weight:
    40 kg
  • Cat. No.:
    735-0390
  • Detector:
    PMT and/or photodiode
  • Wavelength selection:
    Choice of simultaneous detection of all wavelengths or selection in 1,0 nm increments
  • Model:
    SpectraMax® iD3
  • Wavelength range:
    230 - 1000 nm
  • W×D×H:
    532×598×401 mm
  • Plate types:
    6- to 384-well plates
Complementary Products

Product Information

  • Catalog #
    Description
    Pk
    Serum format
    Sterility
    Packed
    Recommended working volume
    Culture area
    Sterile
    Colour
    Availability
    Price
  • 30009-2
    Caspase-3 DEVD-R110 Fluorometric HTS Assay Kit, 10 ml
    1 KIT
    Log in for availability
    1 * 1 KIT - 294,00 €

  • 30026.
    Calcein-AM Cell Viability Assay kit, 1000 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - 417,00 €

  • 30002.
    Viability/Cytotoxicity Assay kit for Animal Live & Dead Cells, 300 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - 440,00 €

  • 30029.
    NucView™ 488 Caspase-3 Assay Kit for live cells, 100 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - 631,00 €

  • 31006.
    AccuBlue® High sensitivity dsDNA quantitation kit with 8 DNA standards
    1.000 Assays
    Log in for availability
    1 * 1.000 Assays - 435,00 €

  • 31007.
    AccuBlue® Broad range dsDNA quantitation kit with 9 DNA standards
    1.500 Assays
    Log in for availability
    1 * 1.500 Assays - 435,00 €

  • S1810-500
    Foetal bovine serum, South American origin
    Frozen
    15
    Log in for availability
    1 * 500 mL - 322,00 €

  • 31028.
    AccuClear® Ultra high sensitivity dsDNA quantitation kit with 7 DNA standards
    1.000 Assays
    Log in for availability
    1 * 1.000 Assays - 518,00 €

  • 734-2327
    1/bag
    0,075 - 0,20 ml
    0,33 cm²
    +
    17
    Clear
    Log in for availability
    1 * 100 unid. - 185,00 €