Entrada de Pedido
Portugal
ContactUsLinkComponent
MTase-Glo™ Methyltransferase Assay The system contains sufficient reagents for 400 assays in 96-well plates, or 2,000 assays in low-volume 384-well plates. Includes: • 300µl SAH, 15µM • 200µl SAM, 1mM • 1ml MTase-Glo™ Reagent, 10X • 10ml MTase-Glo™ D 1 * 400 Tests
Catalog # PROMV7601
Supplier:  Promega
undefined
MTase-Glo™ Methyltransferase Assay The system contains sufficient reagents for 400 assays in 96-well plates, or 2,000 assays in low-volume 384-well plates. Includes: • 300µl SAH, 15µM • 200µl SAM, 1mM • 1ml MTase-Glo™ Reagent, 10X • 10ml MTase-Glo™ D 1 * 400 Tests
Catalog # PROMV7601
Supplier:  Promega
Frequently Bought Together