Order Entry
Portugal
ContactUsLinkComponent
Protein A is a cell-wall receptor which is present on cell wall of various starins of Staphyloccocus aureus. Protein A contains four high-affinity binding sites for interacting with the Fc region of immunoglobulin Gs of various species including mou 1 * 5 mg
Catalog # GENO786-1266
Supplier:  G-Biosciences
Protein A is a cell-wall receptor which is present on cell wall of various starins of Staphyloccocus aureus. Protein A contains four high-affinity binding sites for interacting with the Fc region of immunoglobulin Gs of various species including mou 1 * 5 mg
Catalog # GENO786-1266
Supplier:  G-Biosciences
Frequently Bought Together