Order Entry
Portugal
ContactUsLinkComponent
Sipper Sample Comparment Accessory for GENESYS 140, 150, 180 & and BioMate 160. Sample compartment insert with cuvette holder and g oose-neck aspiration tube guide. Does not include pump, communications cable, power supply or flow cell. 1 * 1 items
Catalog # 634-1110
Sipper Sample Comparment Accessory for GENESYS 140, 150, 180 & and BioMate 160. Sample compartment insert with cuvette holder and g oose-neck aspiration tube guide. Does not include pump, communications cable, power supply or flow cell. 1 * 1 items
Catalog # 634-1110
Frequently Bought Together