Order Entry
Portugal
ContactUsLinkComponent
Replacement Filter cells for 4' Purifier Logic+ and Cell Logic+ Disposable Prefilter Kit. Includes G4 pleated disposable (MERV 8) filter cells. EA 1 * 1 KIT
Catalog # LBCP3406201
Supplier:  LABCONCO
Replacement Filter cells for 4' Purifier Logic+ and Cell Logic+ Disposable Prefilter Kit. Includes G4 pleated disposable (MERV 8) filter cells. EA 1 * 1 KIT
Catalog # LBCP3406201
Supplier:  LABCONCO
Frequently Bought Together