Order Entry
Portugal
ContactUsLinkComponent
Refill-Cylinder-Therm., 0+150:1C, enclosed scale, cap. lens front, colourless, red special liquid filled, c. 250x17mm, cork-glass-t op finish 1 * 1 items
Catalog # AMART108296-FL
Supplier:  Amarell
Refill-Cylinder-Therm., 0+150:1C, enclosed scale, cap. lens front, colourless, red special liquid filled, c. 250x17mm, cork-glass-t op finish 1 * 1 items
Catalog # AMART108296-FL
Supplier:  Amarell
Frequently Bought Together