Order Entry
Portugal
ContactUsLinkComponent
Therm. to set Amarell Universal, +500+1000:5C, 400x9mm, enclosed scale, cap. round, colourless, gallium alloy, total immersion, pin -top finish 1 * 1 items
Catalog # AMARN71026-GA
Supplier:  Amarell
Therm. to set Amarell Universal, +500+1000:5C, 400x9mm, enclosed scale, cap. round, colourless, gallium alloy, total immersion, pin -top finish 1 * 1 items
Catalog # AMARN71026-GA
Supplier:  Amarell
Frequently Bought Together