Order Entry
Portugal
ContactUsLinkComponent
Thermometer similar to IP 91 C, solid stem, white backed, 0+110:1C, red special liquid, durable pigment, suitable for government ve rification 1 * 1 items
Catalog # AMARA341250-FL
Supplier:  Amarell
Thermometer similar to IP 91 C, solid stem, white backed, 0+110:1C, red special liquid, durable pigment, suitable for government ve rification 1 * 1 items
Catalog # AMARA341250-FL
Supplier:  Amarell
Frequently Bought Together