Order Entry
Portugal
ContactUsLinkComponent
Thermometer similar to IP 40 C, solid stem, white backed, 20+120:1C, red special liquid, durable pigment, suitable for government v erification 1 * 1 items
Catalog # AMARA340570-FL
Supplier:  Amarell
Thermometer similar to IP 40 C, solid stem, white backed, 20+120:1C, red special liquid, durable pigment, suitable for government v erification 1 * 1 items
Catalog # AMARA340570-FL
Supplier:  Amarell
Frequently Bought Together